missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHAT Mouse anti-Human, Unconjugated, Clone: 1F7A9, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
Antigen | CHAT |
---|---|
Clone | 1F7A9 |
Concentration | 1000 μg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16809045
|
Proteintech
66816-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16899035
|
Proteintech
66816-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CHAT antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CHAT. This antibody recognizes human, mouse, rat antigen. The CHAT antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
CHAT | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
1103 | |
Primary |
1F7A9 | |
Western Blot, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG2b |