missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One CELLCOAT™ Cell Culture Flasks
Provide enhanced adherence of cells and a cell environment mimicking the in vivo state of the cell. Greiner Bio-One™ CELLCOAT™ Cell Culture Flasks can be used for studies of cellular differentiation, cell-matrix interactions, receptor-ligand binding, gene expression and regulation, embryogenesis and organ development, drug screening and sensitivity assays and culture of primary tumors.
Brand: Greiner Bio-One
This item is not returnable.
View return policy
Description
- Promotion of cell adhesion, proliferation and growth of endothelial cell, hepatocytes, muscle cells, pheochromocytoma cells (PC12) and other cell types
- Shelf life: 24 months at room temperature
- Cell cultivation in serum-free or serum-reduced medium
- Rat tail Collagen Type I
- Increase in isolation and cultivation efficiency
- Ready-to-use products: immediate use, time-saving
- Consistent quality
- Storable at room temperature
Specifications
50 mL | |
Red filter screw cap | |
50 Pack | |
Collagen Type I Protein | |
5 to 10 mL |
Polystyrene | |
Canted | |
50 | |
Yes |