missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11a/Integrin Alpha L Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | CD11a/Integrin Alpha L |
---|---|
Concentration | 187 μg/mL |
Applications | Western Blot, Immunohistochemistry, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16894804
|
Proteintech
15574-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16884804
|
Proteintech
15574-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD11a/Integrin Alpha L antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CD11a/Integrin Alpha L. This antibody recognizes human antigen. The CD11a/Integrin Alpha L antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
CD11a/Integrin Alpha L | |
Western Blot, Immunohistochemistry, ELISA | |
Unconjugated | |
Human | |
IgG |
187 μg/mL | |
Polyclonal | |
Rabbit | |
3683 | |
Primary |