Learn More
CD11a/Integrin Alpha L Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15574-1-AP-150UL
This item is not returnable.
View return policy
Description
The CD11a/Integrin Alpha L antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CD11a/Integrin Alpha L. This antibody recognizes human antigen. The CD11a/Integrin Alpha L antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
CD11a/Integrin Alpha L | |
Polyclonal | |
Unconjugated | |
150 μL | |
3683 | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
187 μg/mL | |
Rabbit | |
Primary | |
Human |