missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC93 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20861-1-AP-20UL
This item is not returnable.
View return policy
Description
CCDC93 is involved in copper-dependent ATP7A trafficking between the trans-Golgi network and vesicles in the cell periphery; the function is proposed to depend on its association within the CCC complex and cooperation with the WASH complex on early endosomes and is dependent on its interaction with WASHC2C.Specifications
CCDC93 | |
Polyclonal | |
Unconjugated | |
CCDC93 | |
CCDC93 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
304743, 54520, 70829 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q567U6, Q5BJT7, Q7TQK5 | |
CCDC93 | |
CCDC93 Fusion Protein Ag14855 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |