missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC28A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14807-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a coiled-coil domain containing protein. Although the specific function of this gene has not yet been determined, this gene is a known translocation partner of nucleoporin 98 in acute leukemias. The resulting fusion gene produces a nucleoporin 98-coiled-coil domain-containing protein 28A chimeric protein which may be involved in promoting myeloproliferative neoplasms.Specifications
CCDC28A | |
Polyclonal | |
Unconjugated | |
CCDC28A | |
C6orf80, CCDC28A, CCRL1AP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
215814, 25901, 361454 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8IWP9 | |
CCDC28A | |
CCDC28A Fusion Protein Ag6529 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |