missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCAR2 Mouse anti-Human, Porcine, Clone: 2D10F8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66497-1-IG-150UL
This item is not returnable.
View return policy
Description
DBC1 is located within a chromosomal region that shows loss of heterozygosity in some bladder cancers. It contains a 5′ CpG island that may be a frequent target of hypermethylation, and it may undergo hypermethylation-based silencing in some bladder cancers.Specifications
CCAR2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8N163 | |
CCAR2 | |
CCAR2 Fusion Protein Ag18374 | |
150 μL | |
Primary | |
Human, Porcine | |
Antibody | |
IgG2a |
Immunocytochemistry, Immunofluorescence, Western Blot | |
2D10F8 | |
Unconjugated | |
CCAR2 | |
CCAR2, cell cycle and apoptosis regulator 2, DBC 1, DBC.1, DBC1, KIAA1967, p30 DBC, Protein KIAA1967 | |
Mouse | |
Protein A | |
RUO | |
100155718, 57805 | |
-20°C | |
Liquid |