missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAPRIN1 Mouse anti-Human, Mouse, Rat, Clone: 6B10F11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66352-1-IG-150UL
This item is not returnable.
View return policy
Description
GPIP137 may regulate the transport and translation of mRNAs of proteins involved in synaptic plasticity in neurons and cell proliferation and migration in multiple cell types. It is expressed in the cytoplasm.Specifications
CAPRIN1 | |
Monoclonal | |
0.74 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q14444, Q5M9G3, Q60865 | |
Caprin1 | |
CAPRIN1 Fusion Protein Ag7413 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
6B10F11 | |
Unconjugated | |
CAPRIN1 | |
caprin 1, CAPRIN1, GPI anchored protein p137, GPI p137, GPIAP1, GPIP137, M11S1, p137GPI, RNG105 | |
Mouse | |
Protein G | |
RUO | |
362173, 4076, 53872 | |
-20°C | |
Liquid |