Learn More
Calcium silicide, tech. Ca ≈30%, may contain up to 5% Fe, Thermo Scientific Chemicals
£41.00 - £173.87
Chemical Identifiers
CAS | 12013-56-8 |
---|---|
Molecular Formula | CaSi2 |
Molecular Weight (g/mol) | 96.248 |
MDL Number | MFCD00015977 |
InChI Key | XXCKHUMTXOCLPT-UHFFFAOYSA-N |
Synonym | calciumdisilicid, ? 1-silylcalcio-? 1-silanylidyne |
PubChem CID | 13848147 |
IUPAC Name | calcium;silicon(1-) |
SMILES | [Si-].[Si-].[Ca+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11366978
|
Thermo Scientific Alfa Aesar
014676.22 |
100 g |
£41.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11376978
|
Thermo Scientific Alfa Aesar
014676.36 |
500 g |
£173.87
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is an important raw material used in Pharmaceutical intermediates. It is an ideal compound of deoxidizer and desulfurizer which can be used in production of many types of stainless steel. It is also used as inoculants and additive in cast iron.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12013-56-8 | |
96.248 | |
XXCKHUMTXOCLPT-UHFFFAOYSA-N | |
13848147 | |
[Si-].[Si-].[Ca+2] |
CaSi2 | |
MFCD00015977 | |
calciumdisilicid, ? 1-silylcalcio-? 1-silanylidyne | |
calcium;silicon(1-) |
Specifications
∼1000°C | |
Gray | |
CaSi2 | |
UN1405 | |
Insoluble in cold water, decomposes in hot water | |
[Si-].[Si-].[Ca+2] | |
96.248 | |
96.25 | |
Ca ≈30%, may contain up to 5% Fe | |
2.5g/mL |
12013-56-8 | |
Powder | |
MFCD00015977 | |
calciumdisilicid, ? 1-silylcalcio-? 1-silanylidyne | |
XXCKHUMTXOCLPT-UHFFFAOYSA-N | |
calcium;silicon(1-) | |
13848147 | |
Technical | |
Moisture Sensitive | |
Calcium silicide |
Safety and Handling
GHS H Statement
H261
In contact with water releases flammable gas.
GHS P Statement
P231+P232-P280-P233-P370+P378c-P402+P404-P501a
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep container tightly closed.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: CALCIUM SILICIDE
missing translation for 'tsca' : TSCA
RUO – Research Use Only