missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadmium Sulfate 8/3-Hydrate, puriss. p.a. ACS Reagent, ≥99.0%, Honeywell Fluka™
Puriss. p.a., ACS Reagent,≥99.0% (calc. based on CdSO4 · 8/3H2O, KT)
£33.28 - £159.33
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15681890
|
Honeywell-Fluka
20920-25G |
25 g | Poly Bottle | N/A | |||||
15671890
|
Honeywell-Fluka
20920-100G |
100 g | Poly Bottle | N/A | |||||
15691890
|
Honeywell-Fluka
20920-500G |
500 g | Poly Bottle | N/A | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7790-84-3 | |
MFCD02253000 |
Specifications
41.5°C | |
Colorless | |
CdSO4 · 8/3H2O | |
UN2570 | |
≥99.0% (calc. based on CdSO4.83H2O, KT) | |
3.09g/cu.cm |
7790-84-3 | |
Solid | |
MFCD02253000 | |
256.57g/mol | |
ACS/puriss. p.a. | |
Cadmium Sulfate 8/3-Hydrate |
Safety and Handling
P201-P260-P301 + P310 + P330-P304 + P340 + P310-P308 + P313-P403 + P233
H301-H330-H340-H350-H360FD-H372-H410
missing translation for 'dotInformation' : class 6.1 - PG 3 - EHS - Cadmium compounds, HI: all (not BR)
missing translation for 'einecsNumber' : 233-331-6