missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Sulfate 8/3-Hydrate, puriss. p.a. ACS Reagent, ≥99.0%, Honeywell Fluka™

Product Code. 15671890
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss. p.a., ACS Reagent,≥99.0% (calc. based on CdSO4 · 8/3H2O, KT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-84-3
MDL Number MFCD02253000

Specifications

Melting Point 41.5°C
Color Colorless
Physical Form Solid
Quantity 100 g
Linear Formula CdSO4 · 8/3H2O
UN Number UN2570
Formula Weight 256.57g/mol
Percent Purity ≥99.0% (calc. based on CdSO4.83H2O, KT)
Grade ACS/puriss. p.a.
Packaging Poly Bottle
Density 3.09g/cu.cm
Chemical Name or Material Cadmium Sulfate 8/3-Hydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'