Learn More
Cadmium acetate dihydrate, 98%, pure, Thermo Scientific Chemicals
£47.74 - £315.57
Chemical Identifiers
CAS | 5743-04-4 |
---|---|
Molecular Formula | C4H6CdO4·2H2O |
Molecular Weight (g/mol) | 266.52 |
InChI Key | AUIZLSZEDUYGDE-UHFFFAOYSA-L |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15249716
|
Thermo Scientific Acros
212312500 |
250 g |
£47.74
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15279666
|
Thermo Scientific Acros
212310010 |
1 kg |
£111.56
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15289666
|
Thermo Scientific Acros
212310050 |
5 kg |
£315.57
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5743-04-4 | |
C4H6CdO4·2H2O | |
266.52 | |
AUIZLSZEDUYGDE-UHFFFAOYSA-L |
Specifications
5743-04-4 | |
C4H6CdO4·2H2O | |
10,1584 | |
AUIZLSZEDUYGDE-UHFFFAOYSA-L | |
266.52 | |
Pure | |
Cadmium acetate dihydrate |
97.5% min. (Complexometry) | |
(CH3CO2)2Cd·2H2O | |
Solubility in water: soluble. Other solubilities: soluble in ethanol | |
266.52 | |
98% | |
12.5% to 14.5% |
Safety and Handling
GHS H Statement
Very toxic to aquatic life with long lasting effects.
May cause cancer.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a posi
GHS Signal Word: Danger
- Cadmium acetate dihydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1A
Hazard Statement
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H350-May cause cancer.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only