missing translation for 'onlineSavingsMsg'
Learn More

CA9 Mouse anti-Human, Mouse, Porcine, Clone: 1G4G5, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66243-1-IG-20UL

 View more versions of this product

Product Code. 16887935

  • £133.90 / 20µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Carbonic anhydrase (CA) is an enzyme that assists rapid interconversion of carbon dioxide and water into carbonic acid, protons, and bicarbonate ions. It is abundant in all mammalian tissues. There are many genes that are inducible by hypoxia, via HIF-1 alpha. CA IX is one of the most inducible genes because of its stability and location within the membrane. Carbonic anhydrases have a widespread role in regulating pH in normal tissues, by regulating hydrogen ion (H+) flux. The pH is important in cell death under hypoxia, thus a blockade of CA IX results in increased cell death under hypoxia. Therefore, CA IX has become a reliable histochemical marker of hypoxia.
Specifications

Specifications

CA9
Monoclonal
1.36 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q16790, Q8VHB5
CA9
CA9 Fusion Protein Ag1540
20 μL
Primary
Mouse, Human, Porcine
Antibody
IgG2a
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1G4G5
Unconjugated
CA9
CA IX, CA9, CAIX, Carbonate dehydratase IX, Carbonic anhydrase 9, carbonic anhydrase IX, G250, Membrane antigen MN, MN, P54/58N, pMW1, RCC associated antigen G250
Mouse
Protein G
RUO
100152792, 230099, 768
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers