missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CA9 Mouse anti-Human, Mouse, Porcine, Clone: 1G4G5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66243-1-IG-150UL
This item is not returnable.
View return policy
Description
Carbonic anhydrase (CA) is an enzyme that assists rapid interconversion of carbon dioxide and water into carbonic acid, protons, and bicarbonate ions. It is abundant in all mammalian tissues. There are many genes that are inducible by hypoxia, via HIF-1 alpha. CA IX is one of the most inducible genes because of its stability and location within the membrane. Carbonic anhydrases have a widespread role in regulating pH in normal tissues, by regulating hydrogen ion (H+) flux. The pH is important in cell death under hypoxia, thus a blockade of CA IX results in increased cell death under hypoxia. Therefore, CA IX has become a reliable histochemical marker of hypoxia.Specifications
CA9 | |
Monoclonal | |
1.36 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q16790, Q8VHB5 | |
CA9 | |
CA9 Fusion Protein Ag1540 | |
150 μL | |
Primary | |
Human, Mouse, Porcine | |
Antibody | |
IgG2a |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1G4G5 | |
Unconjugated | |
CA9 | |
CA IX, CA9, CAIX, Carbonate dehydratase IX, Carbonic anhydrase 9, carbonic anhydrase IX, G250, Membrane antigen MN, MN, P54/58N, pMW1, RCC associated antigen G250 | |
Mouse | |
Protein G | |
RUO | |
100152792, 230099, 768 | |
-20°C | |
Liquid |