missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17239-1-AP-20UL
This item is not returnable.
View return policy
Description
C6 is a component of complement cascade. It is part of the membrane attack complex which can insert into the cell membrane and cause cell to lyse. People with C6 deficiency are prone to bacterial infection. Two transcript variants encoding the same protein have been found for this gene.Specifications
C6 | |
Polyclonal | |
Unconjugated | |
C6 | |
C6, complement component 6, Complement component C6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12274, 24237, 729 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P13671, Q811M5 | |
C6 | |
C6 Fusion Protein Ag11083 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |