missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C3orf43 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25362-1-AP-20UL
This item is not returnable.
View return policy
Description
SMCO1 is a protein coding gene.Specifications
C3orf43 | |
Polyclonal | |
Unconjugated | |
SMCO1 | |
C3orf43 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
255798, 69576 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunofluorescence | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q147U7, Q8CEZ1 | |
Smco1 | |
C3orf43 Fusion Protein Ag21670 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |