Learn More
C20orf20 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26040-1-AP-20UL
 View more versions of this product
Product Code. 16884565
  • 145.00€ / 20µL
Explore more special offers
Description
Description
C20orf20, also named as MRG/MORF4L-binding protein or Up-regulated in colon cancer 4 (Urcc4), is a 204 amino acid protein, which belongs to the EAF7 family. C20orf20 localizes in the nucleus and is a Component of the NuA4 histone acetyltransferase (HAT) complex which is involved in transcriptional activation of select genes principally by acetylation of nucleosomal histones H4 and H2A. This modification may both alter nucleosome - DNA interactions and promote interaction of the modified histones with other proteins which positively regulate transcription. This complex may be required for the activation of transcriptional programs associated with oncogene and proto-oncogene mediated growth induction, tumor suppressor mediated growth arrest and replicative senescence, apoptosis, and DNA repair. C20orf20 is modified through phosphorylation and acetylation, so the molecular weight of C20orf20 might be 25-30 kDa.
Specifications
Specifications
C20orf20
Polyclonal
Unconjugated
MRGBP
C20orf20, Eaf7, MRG binding protein, MRG15BP, MRGBP, URCC4
Rabbit
Antigen Affinity Chromatography
RUO
55257, 73247
-20°C
Liquid
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9DAT2, Q9NV56
MRGBP
C20orf20 Fusion Protein Ag22840
20 μL
Primary
Human, Mouse