missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution, pH 3.00 (+/-0.026 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals

£31.56 - £51.96

Specifications

Chemical Name or Material Buffer solution
CAS 1336-21-6
Color Colorless
Physical Form Liquid
Molecular Formula H5NO
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11324578
View Documents
Thermo Scientific Alfa Aesar
042416.K2
1 L
£31.56
1 L
Please to purchase this item. Need a web account? Register with us today!
11334578
View Documents
Thermo Scientific Alfa Aesar
042416.K7
4 L
£51.96
4 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Buffer solution (pH 3) is used as a means of keeping pH at a nearly constant value in different chemical applications. Buffer solution is used in alcoholic fermentation, leather tanning, electroplating, organic synthesis, sugar and paper manufacturing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Buffer solution
Colorless
H5NO
MFCD00066650
VHUUQVKOLVNVRT-UHFFFAOYSA-N
35.05
CHEBI:18219
1336-21-6
Liquid
Odorless
Miscible with water.
N.O
14923
Videos
Safety and Handling

Safety and Handling

P234-P390

H290

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only