missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 3.60, Chem-Lab

Product Code. 11934034
Click to view available options
Quantity:
10 L
Packaging:
PE
This item is not returnable. View return policy
This item is not returnable. View return policy

Phtalate-HCl buffer for laboratory use and pH measurements

Specifications

Density 1.01g/mL
pH 3.60
Quantity 10 L
Formulation Phtalate-HCl
Packaging PE
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'