missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BSA Mouse anti-Bovine, Clone: 2D11G3, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
Antigen | BSA |
---|---|
Clone | 2D11G3 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16887855
|
Proteintech
66201-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16877855
|
Proteintech
66201-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody recognizes albumin from bovine but not human, rat nor mouse.
Albumin is a soluble, monomeric protein which comprises about one-half of the blood serum protein. Albumin functions primarily as a carrier protein for steroids, fatty acids, and thyroid hormones and plays a role in stabilizing extracellular fluid volume. Albumin is a globular unglycosylated serum protein of molecular weight 65, 000. Albumin is synthesized in the liver as preproalbumin which has an N-terminal peptide that is removed before the nascent protein is released from the rough endoplasmic reticulum. The product, proalbumin, is in turn cleaved in the Golgi vesicles to produce the secreted albumin.Specifications
BSA | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Bos d 6, BSA, Serum albumin | |
ALB | |
IgG1 | |
Protein G | |
Antibody |
2D11G3 | |
Western Blot | |
Unconjugated | |
Mouse | |
Bovine | |
P02769 | |
280717 | |
Peptide | |
Primary | |
-20°C | |
ALB |