missing translation for 'onlineSavingsMsg'
Learn More

BSA Mouse anti-Bovine, Clone: 2D11G3, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66201-1-IG-150UL

 View more versions of this product

Product Code. 16877855

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This antibody recognizes albumin from bovine but not human, rat nor mouse.

Albumin is a soluble, monomeric protein which comprises about one-half of the blood serum protein. Albumin functions primarily as a carrier protein for steroids, fatty acids, and thyroid hormones and plays a role in stabilizing extracellular fluid volume. Albumin is a globular unglycosylated serum protein of molecular weight 65, 000. Albumin is synthesized in the liver as preproalbumin which has an N-terminal peptide that is removed before the nascent protein is released from the rough endoplasmic reticulum. The product, proalbumin, is in turn cleaved in the Golgi vesicles to produce the secreted albumin.
Specifications

Specifications

BSA
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P02769
ALB
Peptide
150 μL
Primary
Bovine
Antibody
IgG1
Western Blot
2D11G3
Unconjugated
ALB
Bos d 6, BSA, Serum albumin
Mouse
Protein G
RUO
280717
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers