Learn More
Brady™ Brandy External Roll Holder
A small and compact desktop printer which provides high quality thermal transfer printing at a low cost
Brand: Brady™ 361065
Code : 50
Additional Details : Weight : 0.10000kg
Description
The Brady™ BBP11 printer is a small and compact desktop printer which provides high quality thermal transfer printing at a low cost. The printer offers high performance with durable and reliable output, with a printout speed of up to 127mm (5inch) per second. It is easy to use and offers quick ribbon and media loading. The standard internal Ethernet port enables easy integration into any application.- Small and compact only 101.6mm (4inch) wide
- 300dpi print resolution
- Offers a wide range of printable materials
- Reliable and durable
- Many features built-in as standard such as Ethernet (network), triple communication interface and dual walled construction Ideal entry level thermal transfer printer for blood, tissue and slide labels in hospitals, healthcare and medical laboratories.
Specifications
External Material Holder for BBP12 and BBP11 | |
1 Ea. |