missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Blocker™ Casein

Block membranes, plates and other samples with these ready-to-use PBS or TBS solutions of purified casein for Western blot, ELISA and other detection methods.

£43.30 - £140.00


Products 4
Product Code Brand Description Quantity Price Quantity & Availability  
Product Code Brand Description Quantity Price Quantity & Availability  
10496134
View Documents
Thermo Scientific™
37528
Blocker Casein in PBS 1 L
£140.00
1 L
View alternative products
Save Up to 
»
In stock, ready to ship: 62
Please to purchase this item. Need a web account? Register with us today!
10443644
View Documents
Thermo Scientific™
37532
Blocker Casein in TBS 1 L
£138.00
1 L
View alternative products
Save Up to 
»
In stock, ready to ship: 5
Please to purchase this item. Need a web account? Register with us today!
13454299
View Documents
Thermo Scientific™
37582
Blocker Casein in PBS 100 mL
£44.23
100mL
View alternative products
Save Up to 
»
In stock, ready to ship: 11
Please to purchase this item. Need a web account? Register with us today!
13464299
View Documents
Thermo Scientific™
37583
Blocker Casein in TBS 100 mL
£43.30
100mL
View alternative products
Save Up to 
»
Estimated Shipment: 01-07-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thermo Scientific™ Blocker Casein Blocking Buffers are ready-to-use, PBS or TBS solutions of purified casein protein for blocking steps in Western blotting, ELISA, immunohistochemistry and nucleic acid detection methods.

These blocking buffers contains casein protein that is purified from milk by the Hammarsten method. The phosphate- and Tris-buffered saline formulations are used for blocking excess binding sites on membranes and microplates in antibody-based detection methods. Blocker Casein Buffers are 1% (w/v) casein, which corresponds to the optimal concentration for most applications.

Highlights:

  • Purified casein – single-protein blocking buffer provides fewer chances of cross-reaction with assay components than serum or milk solutions

  • Easy to use – 1% casein solutions are ready to use; can be diluted further as needed

  • Flexible – available in PBS and TBS formulations to suit a variety of applications

  • Safe – stable, thimerosal-free formulations

SDS
Product Certifications