Learn More
Bismuth(III) nitrate pentahydrate, 98%, pure, Thermo Scientific Chemicals
£103.52 - £436.17
Chemical Identifiers
CAS | 10035-06-0 |
---|---|
Molecular Formula | BiH10N3O14 |
Molecular Weight (g/mol) | 485.07 |
MDL Number | MFCD00149157 |
InChI Key | FBXVOTBTGXARNA-UHFFFAOYSA-N |
Synonym | bismuth nitrate pentahydrate, bismuth trinitrate pentahydrate, bismuth nitrate, hydrate 1:5, bismuth iii nitrate pentahydrate, unii-fo2icy167b, fo2icy167b, nitric acid, bismuth 3+ salt, pentahydrate, acmc-20alle, bismuthnitratepentahydrate |
PubChem CID | 160911 |
IUPAC Name | bismuth;trinitrate;pentahydrate |
SMILES | O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15239916
|
Thermo Scientific Acros
454110010 |
1 kg |
£103.52
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15249916
|
Thermo Scientific Acros
454110050 |
5 kg |
£436.17
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10035-06-0 | |
485.07 | |
FBXVOTBTGXARNA-UHFFFAOYSA-N | |
160911 | |
O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
BiH10N3O14 | |
MFCD00149157 | |
bismuth nitrate pentahydrate, bismuth trinitrate pentahydrate, bismuth nitrate, hydrate 1:5, bismuth iii nitrate pentahydrate, unii-fo2icy167b, fo2icy167b, nitric acid, bismuth 3+ salt, pentahydrate, acmc-20alle, bismuthnitratepentahydrate | |
bismuth;trinitrate;pentahydrate |
Specifications
80.0°C | |
10035-06-0 | |
BiH10N3O14 | |
MFCD00149157 | |
bismuth nitrate pentahydrate, bismuth trinitrate pentahydrate, bismuth nitrate, hydrate 1:5, bismuth iii nitrate pentahydrate, unii-fo2icy167b, fo2icy167b, nitric acid, bismuth 3+ salt, pentahydrate, acmc-20alle, bismuthnitratepentahydrate | |
FBXVOTBTGXARNA-UHFFFAOYSA-N | |
bismuth;trinitrate;pentahydrate | |
160911 | |
98% | |
Bismuth(III) nitrate pentahydrate |
30.0°C | |
97.5% min. (Complexometry) | |
Bi(NO3)3·5H2O | |
15,1271 | |
Solubility in water: may decompose. | |
O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
485.07 | |
485.07 | |
Pure |
Safety and Handling
GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May intensify fire; oxidizer.
GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
GHS Signal Word: Danger
- Bismuth(III) nitrate pentahydrate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H272-May intensify fire; oxidiser.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only