missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Binding Buffer MX

Binding buffer adjusts the binding conditions in order to effectively isolate small and large RNA using compatible NucleoSpin™ RNA purification kits and with the NucleoZOL reagent.

Brand:  Macherey-Nagel™ 740405.60


Product Code. 15809097

  • £77.40 / 60mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Supplemental binding buffer for use with NucleoSpin™ RNA kits: miRNA, totalRNA FFPE
  • Enables use of NucleoZOL reagent with NucleoSpin 96 RNA kits
  • Adjusts binding conditions for binding of small and large RNA to the silica membrane
Specifications

Specifications

Colorless
60 mL
Odorless
Liquid
6.0 to 8.0
123-91-1
<2% in Water
1,4-dioxane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Buffer MX (60 mL)

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

Supplemental information
  • MIXTURE LIST-Contains : 1,4-dioxane

SDS
Documents

Documents

Certificates
Special Offers

Special Offers