missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Alcohol, ReagentPlus™, ≥99%, Honeywell™

Product Code. 15610990
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-51-6
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00004599,MFCD03792087
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
Synonym benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
PubChem CID 244
ChEBI CHEBI:17987
IUPAC Name phenylmethanol
SMILES OCC1=CC=CC=C1

Specifications

Melting Point -15.4°C
Color Colorless
Density 1.045g/mL at 25°C
Boiling Point 205.3°C
Flash Point 96°C (204.8°F)
Ignition Point 817°F
Packaging Stainless Steel Can
Linear Formula C6H5CH2OH
Refractive Index n20/D 1.539(lit.)
Quantity 20 L
UN Number UN3334
Beilstein 878307
Vapor Density 3.7 (vs air)
Formula Weight 108.14g/mol
Vapor Pressure 3.75mmHg (77°C), 13.3mmHg (100°C)
Percent Purity ≥99%
Grade ReagentPlus™
Physical Form Liquid
Chemical Name or Material Benzyl Alcohol
Show More Show Less
compliance-icons
Product Identifier
  • Benzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302 + H332-Harmful if swallowed or if inhaled.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'