Learn More
Bel-Art™ SP Scienceware™ Poxygrid 60-Place Test Tube Racks
To organize and process high volumes of samples.
Brand: Bel-Art™ F18763-0160
Code : 04
Additional Details : Weight : 1.00000kg
Description
- Steel wire test tube rack
- Chemical and corrosion-resistant
- Coated with heat-cured epoxy
- Unaffected by organic solvents, salts, weak acids and weak alkalis
- Temperature range: −223°C (−369°F) to 121°C (250°F)
- Autoclavable
- With square holes, three decks, slotted grid bottom and reinforced edges