missing translation for 'onlineSavingsMsg'
Learn More

BCL11B Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen BCL11B
Concentration 0.4 mg/mL
Applications Immunoprecipitation, Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16896875
View Documents
Proteintech
55414-1-AP-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16886875
View Documents
Proteintech
55414-1-AP-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody is specific to BCL11B. The calculated molecular weight of BCL11B is 95 kDa, but modified BCL11B protein is about 116 kDa.

This gene encodes a C2H2-type zinc finger protein and is closely related to BCL11A, a gene whose translocation may be associated with B-cell maligcies. The specific function of this gene has not yet been determined. Two alternatively spliced transcript variants, which encode distinct isoforms, have been reported.
Specifications

Specifications

BCL11B
Immunoprecipitation, Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
Rabbit
Human, Mouse
Q99PV8, Q9C0K0
58208, 64919
Peptide
Primary
-20°C
BCL11B
0.4 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
B cell CLL/lymphoma 11B, B cell lymphoma/leukemia 11B, BCL11B, COUP TF interacting protein 2, CTIP 2, CTIP2, hRit1, hRIT1 alpha, RIT1
BCL11B
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications