Learn More
BCL11B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 55414-1-AP-150UL
 View more versions of this product
Product Code. 16886875
  • £349.00 / 150µL
Explore more special offers
Description
Description
This antibody is specific to BCL11B. The calculated molecular weight of BCL11B is 95 kDa, but modified BCL11B protein is about 116 kDa.
This gene encodes a C2H2-type zinc finger protein and is closely related to BCL11A, a gene whose translocation may be associated with B-cell maligcies. The specific function of this gene has not yet been determined. Two alternatively spliced transcript variants, which encode distinct isoforms, have been reported.
Specifications
Specifications
BCL11B
Polyclonal
Unconjugated
BCL11B
B cell CLL/lymphoma 11B, B cell lymphoma/leukemia 11B, BCL11B, COUP TF interacting protein 2, CTIP 2, CTIP2, hRit1, hRIT1 alpha, RIT1
Rabbit
Antigen Affinity Chromatography
RUO
58208, 64919
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q99PV8, Q9C0K0
BCL11B
Peptide
150 μL
Primary
Human, Mouse