missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady Printer BBP33 EU, MW software 101.6mm max. label width 230.6mm x 241.6mm x 304.8mm, Brady
Brand: Brady 711073
Printer BBP33 EU, MW software 101.6mm max. label width 230.6mm x 241.6mm x 304.8mm, Brady
Code : 04
Additional Details : Weight : 5.30000kg