missing translation for 'onlineSavingsMsg'
Learn More

Barium Sulfate, 99%, ReagentPlus™, Honeywell™

Product Code. 15682600
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-43-7
Molecular Formula BaO4S
Molecular Weight (g/mol) 233.38
MDL Number MFCD00003455
InChI Key TZCXTZWJZNENPQ-UHFFFAOYSA-L
Synonym barium sulfate, barite, baritop, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast
PubChem CID 24414
SMILES [Ba++].[O-]S([O-])(=O)=O

Specifications

Melting Point 1580°C
Color White
Quantity 1 kg
Linear Formula BaSO4
UN Number NONH for all modes of transport
IUPAC Name barium(2+);sulfate
Formula Weight 233.39g/mol
Percent Purity 99%
Grade ReagentPlus™
Packaging Poly Bottle
Appearance Powder
Density 4.4g/cm3
Chemical Name or Material Barium Sulfate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'