missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Bulkhead Compression Union, PVDF

Cole-Parmer™ Bulkhead Compression Union, PVDF has very good chemical and thermal resistance.

Brand:  Cole-Parmer™ 06390-50

Product Code. 11774819

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Very good chemical resistance.
  • Rated to 220psi at 77°F (25°C).
  • Usable from 32 to 212°F (0 to 100°C).
Specifications

Specifications

Autoclavable
1/4 in. OD tubing
7/32 in. orifice; 2-9/32 x 5/8 in.
PVDF
1 Each
Autoclave or Ethylene Oxide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers