missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP5Z1/SPG48 Mouse anti-Human, Rat, Clone: 1G1D7, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
Antigen | AP5Z1/SPG48 |
---|---|
Clone | 1G1D7 |
Concentration | 0.7 mg/mL |
Applications | Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16888495
|
Proteintech
66533-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16878495
|
Proteintech
66533-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The function of this protein has not been specifically defined.Specifications
AP5Z1/SPG48 | |
0.7 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
AP-5 complex subunit zeta-1, AP5Z1, KIAA0415, SPG48, Zeta5 | |
Ap5z1 | |
IgG1 | |
Protein G | |
Antibody |
1G1D7 | |
Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human, Rat | |
O43299 | |
641386, 9907 | |
AP5Z1/SPG48 Fusion Protein Ag26961 | |
Primary | |
-20°C | |
AP5Z1 |