missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP5Z1/SPG48 Mouse anti-Human, Rat, Clone: 1G1D7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66533-1-IG-20UL
This item is not returnable.
View return policy
Description
The function of this protein has not been specifically defined.Specifications
AP5Z1/SPG48 | |
Monoclonal | |
0.7 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43299 | |
Ap5z1 | |
AP5Z1/SPG48 Fusion Protein Ag26961 | |
20 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1G1D7 | |
Unconjugated | |
AP5Z1 | |
AP-5 complex subunit zeta-1, AP5Z1, KIAA0415, SPG48, Zeta5 | |
Mouse | |
Protein G | |
RUO | |
641386, 9907 | |
-20°C | |
Liquid |