missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP3S2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15319-1-AP-150UL
This item is not returnable.
View return policy
Description
AP3S2 (Adaptor Related Protein Complex 3 Subunit Sigma 2) is a Protein Coding gene. Among its related pathways are Lysosome. Gene Ontology (GO) annotations related to this gene include protein transporter activity. An important paralog of this gene is AP3S1. Part of the AP-3 complex, an adaptor-related complex which is not clathrin-associated. The complex is associated with the Golgi region as well as more peripheral structures. It facilitates the budding of vesicles from the Golgi membrane and may be directly involved in trafficking to lysosomes. In concert with the BLOC-1 complex, AP-3 is required to target cargos into vesicles assembled at cell bodies for delivery into neurites and nerve terminals.Specifications
AP3S2 | |
Polyclonal | |
Unconjugated | |
AP3S2 | |
AP 3 complex subunit sigma 2, AP 3 complex subunit sigma 3B, AP3S2, AP3S3, Sigma 3B adaptin, Sigma adaptin 3b, sigma3b, Sigma3B adaptin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10239, 11778, 683402 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P59780, Q8BSZ2 | |
Ap3s2 | |
AP3S2 Fusion Protein Ag7565 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |