Learn More
WTAP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527764
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Jurkat nuclear extract, K562 nuclear extract. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (90%), Xenopus laevis (92%), Pig (100%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
WTAP Polyclonal antibody specifically detects WTAP in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
WTAP | |
Polyclonal | |
Unconjugated | |
WTAP | |
2810408K05Rik; 9430038B09Rik; Female-lethal(2)D homolog; hFL(2)D; KIAA0105; Mum2; PNAS-132; pre-mRNA-splicing regulator WTAP; putative pre-mRNA splicing regulator female-lethal(2D); RGD1563824; Wilms tumor 1 associated protein; wilms tumor 1-associating protein; Wilms' tumour 1-associating protein; Wilms tumour 1-associating protein; WT1-associated protein; WTAP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9589 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q15007 | |
WTAP | |
Recombinant fragment corresponding to a region within amino acids 1 and 215 of Human WTAP | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |