Learn More
WNT9B Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA548036
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In Western blots, less than 5% cross-reactivity with recombinant human (rh) Wnt-2, rhWnt-9a, recombinant mouse (rm) Wnt-1, rmWnt-2b, rmWnt-3a, rmWnt-4, rmWnt-5a, rmWnt-5b, rmWnt-11, and rmWnt-16 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
WNT9B Polyclonal antibody specifically detects WNT9B in Mouse samples. It is validated for Immunohistochemistry (Frozen), Western Blot.Specifications
WNT9B | |
Polyclonal | |
Unconjugated | |
Wnt9b | |
clf; clf1; protein Wnt-14b; Protein Wnt-15; Protein Wnt-9b; UNQ6973/PRO21956; wingless-type MMTV integration site 15; wingless-type MMTV integration site 9B; wingless-type MMTV integration site family member 9B; wingless-type MMTV integration site family, member 15; wingless-type MMTV integration site family, member 9B; Wnt family member 9B; Wnt14b; wnt-14b; Wnt15; wnt-15; Wnt9b | |
Goat | |
Antigen affinity chromatography | |
RUO | |
22412 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Frozen), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O35468 | |
Wnt9b | |
E. coli-derived recombinant mouse Wnt-9b Ala46-Glu90, Lys228-Tyr298 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |