missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WNT10B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA520530
Code : LD
Additional Details : Weight : 0.01000kg
Description
A suggested positive control is human skeletal muscle tissue lysate. PA5-20530 can be used with blocking peptide PEP-0650.
WNT10B Polyclonal antibody specifically detects WNT10B in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
WNT10B | |
Polyclonal | |
Unconjugated | |
WNT10B | |
Protein Wnt-10b; Protein Wnt-12; SHFM6; wingless related MMTV integration site 10b; wingless-type MMTV integration site family member 10B; wingless-type MMTV integration site family, member 10B; Wnt family member 10B; Wnt10; WNT10B; Wnt-10b; WNT-10B protein; WNT12; wnt-12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22410, 315294, 7480 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O00744, P48614 | |
WNT10B | |
A 15 amino acid peptide from near the center of human Wnt10b. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |