missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WIPI1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534452
Code : NEW
Additional Details : Weight : 0.01000kg
Description
A suggested positive control is rat colon lysate. PA5-34452 can be used with blocking peptide PEP-1494.
WIPI1 Polyclonal antibody specifically detects WIPI1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
WIPI1 | |
Polyclonal | |
Unconjugated | |
WIPI1 | |
4930533H01Rik; ATG18; Atg18 protein homolog; ATG18A; AW411817; D11Ertd498e; RGD1307754; WD repeat domain phosphoinositide-interacting protein 1; WD repeat domain, phosphoinositide interacting 1; WD40 repeat protein interacting with phosphoinositides of 49 kDa; WD40 repeat protein Interacting with phosphoInositides of 49kDa; WIPI 49 kDa; Wipi1; WIPI-1; WIPI-1 alpha; WIPI49 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
303630, 52639, 55062 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q5MNZ9, Q8R3E3 | |
WIPI1 | |
A 17 amino acid synthetic peptide near the carboxy terminus of human WIPI1 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |