missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WFDC5 Monoclonal Antibody (5G5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA523092
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide Sequence: RKKGEKSGGC PPDDGPCLLS VPDQCVEDSQ CPLTRKCCYR ACFRQCVPRV SVKLGSCPED QLRCLSPMNH LCHKDSDCSG KKRCCHSACG RDCRDPARG
WFDC5 Monoclonal antibody specifically detects WFDC5 in Human samples. It is validated for ELISA, Western Blot.Specifications
WFDC5 | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q8TCV5 | |
WFDC5 | |
WFDC5 (NP_663627, 25 a.a. ∽ 123 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
5G5 | |
Unconjugated | |
WFDC5 | |
BC019734; dJ211D12.5; p53-responsive gene 5 protein; Prg5; protease inhibitor WAP1; putative protease inhibitor WAP1; UNQ3036/PRO9842; WAP four-disulfide core domain 5; WAP four-disulfide core domain protein 5; WAP1; WFDC5 | |
Mouse | |
Affinity chromatography | |
RUO | |
149708 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |