missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WFDC1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA537972
Code : NEW
Additional Details : Weight : 0.00900kg
Description
This antibody is predicted to react with rat based on sequence homology.
WFDC1 Polyclonal antibody specifically detects WFDC1 in Mouse samples. It is validated for Western Blot.Specifications
WFDC1 | |
Polyclonal | |
Unconjugated | |
WFDC1 | |
2310058A03Rik; Prostate stromal protein ps20; PS20; ps20 growth inhibitor; WAP four-disulfide core domain 1; WAP four-disulfide core domain 1 homolog; WAP four-disulfide core domain protein 1; Wfdc1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
67866 | |
-20°C | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9ESH5 | |
WFDC1 | |
Peptide with sequence C-RQRLHKEYPEGDSKN, from the internal region near the N-terminus of mouse Wfdc1 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |