missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541981
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: LAGHTKAVSS VKFSPNGEWL ASSSADKLIK IWGAYDGKFE KTISGHKLGI Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Yeast: 91%; Zebrafish: 93%
WDR5 Polyclonal antibody specifically detects WDR5 in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blot.Specifications
WDR5 | |
Polyclonal | |
Unconjugated | |
WDR5 | |
2410008O07Rik; AA408785; AA960360; Big; BIG3; BIG-3; BMP2-induced 3-kb gene protein; CFAP89; cilia and flagella associated protein 89; SWD3; SWD3, Set1c WD40 repeat protein, homolog; Unknown (protein for MGC:137327); WD repeat domain 5; WD repeat-containing protein 5; WD repeat-containing protein BIG-3; Wdr5; wu:fk47f04; zgc:56591; zgc:76895; zgc:76895 protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
11091 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P61964 | |
WDR5 | |
Synthetic peptide directed towards the C-terminal of human WDR5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |