missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR32 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA524133
Code : LD
Additional Details : Weight : 0.01000kg
Description
WDR32 Polyclonal antibody specifically detects WDR32 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
WDR32 | |
Polyclonal | |
Unconjugated | |
DCAF10 | |
AA959934; DCAF10; DDB1 and CUL4 associated factor 10; DDB1- and CUL4-associated factor 10; RGD1304730; WD repeat domain 32; WD repeat-containing protein 32; WDR32 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
79269 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q5QP82 | |
DCAF10 | |
KLH conjugated synthetic peptide between 113-142 amino acids from the N-terminal region of human WDR32 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |