Learn More
WBP11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542840
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: GRRSTSSTKS GKFMNPTDQA RKEARKRELK KNKKQRMMVR AAVLKMKDPK Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%
WBP11 Polyclonal antibody specifically detects WBP11 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
WBP11 | |
Polyclonal | |
Unconjugated | |
WBP11 | |
2510026P17Rik; D6Wsu113e; DKFZp779M1063; hypothetical protein LOC402950; Npw38-binding protein; Npw38-binding protein NpwBP; NpwBP; PPP1R165; protein phosphatase 1, regulatory subunit 165; SH3 domain-binding protein; SH3 domain-binding protein SNP70; SIPP1; SNP70; Splicing factor that interacts with PQBP-1 and PP1; splicing factor, PQBP1 and PP1 interacting; Wbp11; WBP-11; WW domain binding protein 11; WW domain-binding protein 11; zgc:77390 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51729, 60321 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q923D5, Q9Y2W2 | |
WBP11 | |
Synthetic peptide directed towards the N-terminal of human WBP11 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |