missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UTY Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA526082
Code : LD
Additional Details : Weight : 0.01000kg
Description
UTY Polyclonal antibody specifically detects UTY in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.Specifications
UTY | |
Polyclonal | |
Unconjugated | |
UTY | |
histone demethylase UTY; Hydb; Kdm6a; KDM6AL; KDM6C; lysine (K)-specific demethylase 6A; lysine-specific demethylase 6A; male-specific histocompatibility antigen H-YDB; mKIAA4057; RGD1565481; ubiquitous TPR motif protein UTY; ubiquitously transcribed tetratricopeptide repeat containing, Y-linked; ubiquitously transcribed tetratricopeptide repeat gene, X chromosome; ubiquitously transcribed tetratricopeptide repeat gene, Y chromosome; ubiquitously transcribed tetratricopeptide repeat gene, Y-linked; ubiquitously transcribed tetratricopeptide repeat, X chromosome; ubiquitously transcribed TPR gene on Y chromosome; ubiquitously transcribed TPR protein on the Y chromosome; ubiquitously transcribed Y chromosome tetratricopeptide repeat protein; ubiquitously-transcribed TPR protein on the Y chromosome; Ubiquitously-transcribed Y chromosome tetratricopeptide repeat protein; Utx; Uty; UTY1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
7404 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Western Blot | |
0.44 mg/mL | |
PBS with 0.09% sodium azide | |
O14607 | |
UTY | |
KLH conjugated synthetic peptide between 1247-1276 amino acids from the C-terminal region of human UTY | |
400 μL | |
Primary | |
Human | |
Antibody | |
Ig |