Learn More
USP20 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533102
Code : PN
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Marmoset (100%); Orangutan, Gibbon, Rabbit (94%); Hamster (89%); Mouse, Bovine, Bat, Elephant, Panda, Horse, Pig, Opossum, Turkey, Chicken (83%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
USP20 Polyclonal antibody specifically detects USP20 in Human, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin).Specifications
USP20 | |
Polyclonal | |
Unconjugated | |
USP20 | |
1700055M05Rik; AI467231; Deubiquitinating enzyme 20; hVDU2; KIAA1003; LSFR3A; pVHL-interacting deubiquitinating enzyme 2; ubiquitin carboxyl-terminal hydrolase 20; ubiquitin specific peptidase 20; ubiquitin specific protease 20; ubiquitin thioesterase 20; ubiquitin thiolesterase 20; ubiquitin-specific-processing protease 20; Usp20; Vdu2; VHL-interacting deubiquitinating enzyme 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10868 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9Y2K6 | |
USP20 | |
Synthetic 18 amino acid peptide from internal region of human USP20. | |
50 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |