Learn More
UPK3A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532636
Code : PN
Additional Details : Weight : 0.01000kg
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10-20 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is bladder carcinoma.
UPK3A Polyclonal antibody specifically detects UPK3A in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
UPK3A | |
Polyclonal | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
O75631 | |
Upk3a | |
Synthetic peptide corresponding to C-terminus of human uroplakin III. | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Upk3a | |
1110017C07Rik; proplakin IIIa; UP3a; UPIII; UPIIIA; Upk3; UPK3A; uroplakin 3; uroplakin 3A; uroplakin III; Uroplakin-3a | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7380 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |