missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Urocortin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538248
Code : NEW
Additional Details : Weight : 0.00900kg
Description
Urocortin (UCN), a 40 amino acids neuropeptide widely distributed in the central nervous system (CNS), belongs to a family of structurally related peptides including corticotrophin releasing factor (CRF), sauvagine and urotensin I. Urocortin is derived from a larger precursor protein (pro-urocortin, also named corticotensin). Urocortin has biological actions similar to CRF such as stress-related endocrine, autonomic and behavioral responses in mammalian species. It is a potent stimulator of adrenocorticotropic hormone (ACTH) secretion by corticotroph cells from the anterior pituitary in vitro and in vivo.Specifications
Urocortin | |
Polyclonal | |
Unconjugated | |
UCN | |
UI, UROC, urocortin, preproprotein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22226, 29151, 7349 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P55089, P81615, P55090 | |
UCN | |
A synthetic peptide derived from the C-terminal region of human urocortin | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |