missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ULK4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538038
Code : NEW
Additional Details : Weight : 0.00900kg
Description
A recommended positive control is NIH-3T3 cell lysate. Multiple isoforms of ULK4 are known to exist.
ULK4 Polyclonal antibody specifically detects ULK4 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
ULK4 | |
Polyclonal | |
Unconjugated | |
ULK4 | |
4932415A06Rik; A730098P15; FAM7C1; REC01035; RGD1565726; serine/threonine-protein kinase ULK4; ULK4; unc-51 like kinase 4; unc-51-like kinase 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
209012, 54986 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q3V129, Q96C45 | |
ULK4 | |
16 amino acid peptide near the carboxy terminus of human ULK4 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |