missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UGT1A4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542934
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: VVLTPEVNMH IKEEKFFTLT AYAVPWTQKE FDRVTLGYTQ GFFETEHLLK Sequence homology: Human: 100%
UGT1A4 Polyclonal antibody specifically detects UGT1A4 in Human samples. It is validated for Western Blot.Specifications
UGT1A4 | |
Polyclonal | |
Unconjugated | |
UGT1A4 | |
bilirubin UDP-glucuronosyltransferase isozyme 2; bilirubin-specific UDPGT isozyme 2; GNT1; hUG-BR2; UDP glucuronosyltransferase 1 family, polypeptide A4; UDP glucuronosyltransferase family 1 member A4; UDP glycosyltransferase 1 family, polypeptide A4; UDP-glucuronosyltransferase 1 family polypeptide A4s; UDP-glucuronosyltransferase 1-4; UDP-glucuronosyltransferase 1A4; UDP-glucuronosyltransferase 1-D; UDPGT; UDPGT 1-4; UGT1; UGT1.4; UGT1-04; UGT1A4; UGT1A4S; UGT1D; UGT-1D | |
Rabbit | |
Affinity Chromatography | |
RUO | |
54657 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P22310 | |
UGT1A4 | |
Synthetic peptide directed towards the N-terminal of human UGT1A4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |